Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 2 - 3 | 109 | 97 | |
NM_022516.5 | NP_071961.2 | 2 - 3 | 109 | 97 | ||
GenBank mRNA | BC061858.1 | - | 2 - 3 | 104 | 97 | |
CB772933.1 | - | 2 - 3 | 119 | 97 | ||
FN803753.1 | - | 2 - 3 | 79 | 97 | ||
X60789.1 | - | 2 - 3 | 72 | 97 | ||
X74565.1 | - | 2 - 3 | 90 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
