Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 5 - 6 | 669 | 108 | |
NM_022516.5 | NP_071961.2 | 5 - 6 | 669 | 108 | ||
GenBank mRNA | BC061858.1 | - | 5 - 6 | 664 | 108 | |
CK365398.1 | - | 2 - 3 | 255 | 108 | ||
X60789.1 | - | 5 - 6 | 632 | 108 | ||
X60790.1 | - | 1 - 2 | 43 | 108 | ||
X74565.1 | - | 5 - 6 | 650 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
