Assay Details
Gene Symbol: Map2k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133283.1 | NP_579817.1 | 3 - 4 | 479 | 99 | |
XM_006240987.2 | XP_006241049.1 | 3 - 4 | 666 | 99 | ||
GenBank mRNA | BC126084.1 | - | 3 - 4 | 577 | 99 | |
D14592.1 | - | 3 - 4 | 450 | 99 | ||
L14936.1 | - | 3 - 4 | 479 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
