Assay Details
Gene Symbol: Map2k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133283.1 | NP_579817.1 | 7 - 8 | 951 | 155 | |
GenBank mRNA | BC126084.1 | - | 7 - 8 | 1049 | 155 | |
D14592.1 | - | 7 - 8 | 922 | 155 | ||
L14936.1 | - | 7 - 8 | 951 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
