Assay Details
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012114.2 | NP_001012114.2 | 15 - 16 | 3645 | 83 | |
GenBank mRNA | BC083596.1 | - | 11 - 12 | 1640 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P209419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134300 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P209419 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P209419 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P209419 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P211893 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P333329 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10893795 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893787 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893788 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893791 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893783 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893779 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893781 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893780 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893790 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893792 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893778 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893793 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893786 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893782 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893794 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893796 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893785 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893784 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893789 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893777 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10893776 | - | - |
Rat Genome 230 2.0 Array | 1378917_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372738_at | - | - |
Rat Genome 230 2.0 Array | 1378917_at | - | - |
Rat Genome 230 2.0 Array | 1372738_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375372_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375372_at | - | - |
Affymetrix HT Rat Focus Array | 1375372_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378917_at | - | - |
Affymetrix HT Rat Focus Array | 1372738_at | - | - |
Affymetrix HT Rat Focus Array | 1372738_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375372_at | - | - |
Affymetrix HT Rat Focus Array | 1378917_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700001702.rn.1 | TC0700000196.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700001703.rn.1 | TC0700000196.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700001701.rn.1 | TC0700000196.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700001704.rn.1 | TC0700000196.rn.1 | RTA TC |
Rat Expression Set 230 | 1372738_at | - | - |
Rat Expression Set 230 | 1375372_at | - | - |
Rat Expression Set 230 | 1375372_PM_at | - | - |
Rat Expression Set 230 | 1372738_PM_at | - | - |
Rat Expression Set 230 | 1378917_at | - | - |
Rat Expression Set 230 | 1378917_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10893776 | - | - |
Rat Genome U34 Set | rc_AI043962_at | - | - |
Rat Genome U34 Set | rc_AI071194_at | - | - |
Rat Genome U34 Set | rc_AI236603_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21479707 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365554 | - | - |
