Assay Details
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012114.2 | NP_001012114.2 | 6 - 7 | 2600 | 77 | |
GenBank mRNA | BC083596.1 | - | 2 - 3 | 595 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
