Assay Details
Gene Symbol: Mum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108736.2 | NP_001102206.2 | 2 - 3 | 136 | 134 | |
XM_006240973.2 | XP_006241035.1 | 2 - 3 | 169 | 134 | ||
XM_006240974.2 | XP_006241036.1 | 2 - 3 | 202 | 134 | ||
XM_006240975.2 | XP_006241037.1 | 2 - 3 | 1123 | 134 | ||
GenBank mRNA | BC160828.1 | - | 2 - 3 | 136 | 134 | |
CV126168.1 | - | 2 - 3 | 162 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
