Assay Details
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241010.1 | XP_006241072.1 | 1 - 2 | 155 | 97 | |
XM_006241011.1 | XP_006241073.1 | 1 - 2 | 182 | 97 | ||
GenBank mRNA | FQ229401.1 | - | 1 - 2 | 182 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241010.1, XM_006241011.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
