Assay Details
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109614.1 | NP_001103084.1 | 5 - 6 | 640 | 67 | |
XM_006241011.1 | XP_006241073.1 | 5 - 6 | 687 | 67 | ||
GenBank mRNA | BC158787.1 | - | 5 - 6 | 598 | 7304442 | 67 |
FQ220392.1 | - | 5 - 6 | 570 | 67 | ||
FQ229401.1 | - | 5 - 6 | 687 | 67 | ||
FQ230290.1 | - | 5 - 6 | 575 | 67 | ||
FQ234550.1 | - | 5 - 6 | 620 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
