Assay Details
Gene Symbol: Lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241035.2 | XP_006241097.1 | 8 - 9 | 2622 | 115 | |
XM_008765148.1 | XP_008763370.1 | 12 - 13 | 2623 | 115 | ||
XM_008765149.1 | XP_008763371.1 | 12 - 13 | 2894 | 115 | ||
XM_216850.8 | XP_216850.6 | 11 - 12 | 3035 | 115 | ||
GenBank mRNA | FQ212316.1 | - | 11 - 12 | 1859 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
