Assay Details
Gene Symbol: Lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765148.1 | XP_008763370.1 | 5 - 6 | 1491 | 83 | |
XM_008765149.1 | XP_008763371.1 | 5 - 6 | 1762 | 83 | ||
XM_216850.8 | XP_216850.6 | 4 - 5 | 1903 | 83 | ||
GenBank mRNA | FQ212316.1 | - | 4 - 5 | 727 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
