Assay Details
Gene Symbol: Lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241035.2 | XP_006241097.1 | 7 - 8 | 2518 | 65 | |
XM_008765148.1 | XP_008763370.1 | 11 - 12 | 2519 | 65 | ||
XM_008765149.1 | XP_008763371.1 | 11 - 12 | 2790 | 65 | ||
XM_216850.8 | XP_216850.6 | 10 - 11 | 2931 | 65 | ||
GenBank mRNA | FQ212316.1 | - | 10 - 11 | 1755 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
