Assay Details
Gene Symbol: Maged2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271110.1 | NP_001258039.1 | 1 - 2 | 74 | 71 | |
GenBank mRNA | AY724507.1 | - | 1 - 2 | 59 | 71 | |
FM059132.1 | - | 1 - 2 | 74 | 71 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001271110.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
