Assay Details
Gene Symbol: Tpd52l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044295.1 | NP_001037760.1 | 2 - 3 | 241 | 119 | |
XM_006227737.2 | XP_006227799.1 | 2 - 3 | 334 | 119 | ||
XM_006227739.2 | XP_006227801.1 | 2 - 3 | 334 | 119 | ||
XM_008758674.1 | XP_008756896.1 | 2 - 3 | 333 | 119 | ||
GenBank mRNA | BC099809.1 | - | 2 - 3 | 241 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
