Assay Details
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039722.2 | NP_001034811.1 | 4 - 5 | 709 | 119 | |
GenBank mRNA | AJ890206.1 | - | 4 - 5 | 781 | 119 | |
EF474455.1 | - | 4 - 5 | 742 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
