Assay Details
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039722.2 | NP_001034811.1 | 9 - 10 | 1420 | 61 | |
GenBank mRNA | AJ890206.1 | - | 9 - 10 | 1492 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
