Assay Details
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039722.2 | NP_001034811.1 | 2 - 3 | 383 | 73 | |
GenBank mRNA | AJ890206.1 | - | 2 - 3 | 455 | 73 | |
EF474455.1 | - | 2 - 3 | 416 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
