Assay Details
Gene Symbol: Prkar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033679.1 | NP_001028851.2 | 2 - 3 | 346 | 78 | |
XM_008768942.1 | XP_008767164.1 | 2 - 3 | 439 | 78 | ||
GenBank mRNA | BC104679.1 | - | 2 - 3 | 346 | 78 | |
FQ212815.1 | - | 1 - 2 | 454 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
