Assay Details
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191837.2 | NP_001178766.1 | 2 - 3 | 565 | 74 | |
XM_008759400.1 | XP_008757622.1 | 1 - 2 | 59 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191837.2, XM_008759400.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
