Assay Details
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191837.2 | NP_001178766.1 | 3 - 4 | 686 | 68 | |
XM_006229310.2 | XP_006229372.1 | 2 - 3 | 1531 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191837.2, XM_006229310.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
