Assay Details
Gene Symbol: Gabrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024370.3 | NP_077346.3 | 5 - 6 | 692 | 132 | |
GenBank mRNA | M81142.1 | - | 5 - 6 | 674 | 132 | |
X63324.1 | - | 748 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
