Assay Details
Gene Symbol: Plekha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109235.2 | NP_001102705.1 | 12 - 13 | 1466 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P485243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104622 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P485243 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P485243 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P485243 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P485243 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73122 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43523 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14065 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10855661 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855656 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855657 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855655 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855651 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855654 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10855650 | - | - |
Rat Genome 230 2.0 Array | 1377446_PM_at | - | - |
Rat Genome 230 2.0 Array | 1395976_at | - | - |
Rat Genome 230 2.0 Array | 1377446_at | - | - |
Rat Genome 230 2.0 Array | 1395976_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377446_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377446_at | - | - |
Affymetrix HT Rat Focus Array | 1390395_at | - | - |
Affymetrix HT Rat Focus Array | 1395976_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390395_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395976_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400006817.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006819.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006820.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006815.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006818.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006812.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006814.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006816.rn.1 | TC0400000957.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400006811.rn.1 | TC0400000957.rn.1 | RTA TC |
Rat Expression Set 230 | 1377446_PM_at | - | - |
Rat Expression Set 230 | 1377446_at | - | - |
Rat Expression Set 230 | 1395976_PM_at | - | - |
Rat Expression Set 230 | 1395976_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10855650 | - | - |
Rat Genome U34 Set | rc_AI177017_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21238658 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374765 | - | - |
