Assay Details
Gene Symbol: Plekha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109235.2 | NP_001102705.1 | 2 - 3 | 331 | 140 | |
XM_006236506.2 | XP_006236568.1 | 2 - 3 | 416 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
