Assay Details
Gene Symbol: Ghrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012850.1 | NP_036982.1 | 7 - 8 | 778 | 109 | |
XM_008762872.1 | XP_008761094.1 | 7 - 8 | 2141 | 109 | ||
GenBank mRNA | AF122055.1 | - | 7 - 8 | 751 | 109 | |
L01407.1 | - | 7 - 8 | 778 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
