Assay Details
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749741.3 | XP_003749789.1 | 1 - 2 | 447 | 95 | |
GenBank mRNA | AY724479.1 | - | 1 - 2 | 411 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
