Assay Details
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749741.3 | XP_003749789.1 | 6 - 7 | 960 | 67 | |
XM_006236474.2 | XP_006236536.1 | 6 - 7 | 834 | 67 | ||
XM_006236475.2 | XP_006236537.1 | 4 - 5 | 771 | 67 | ||
XM_008762933.1 | XP_008761155.1 | 6 - 7 | 595 | 67 | ||
GenBank mRNA | AY724479.1 | - | 6 - 7 | 924 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
