Assay Details
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749741.3 | XP_003749789.1 | 8 - 9 | 1219 | 63 | |
XM_006236474.2 | XP_006236536.1 | 8 - 9 | 1093 | 63 | ||
XM_006236475.2 | XP_006236537.1 | 6 - 7 | 1030 | 63 | ||
XM_008762933.1 | XP_008761155.1 | 8 - 9 | 854 | 63 | ||
GenBank mRNA | AY724479.1 | - | 8 - 9 | 1183 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
