Assay Details
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107863.1 | NP_001101333.1 | 2 - 3 | 730 | 112 | |
XM_006236571.2 | XP_006236633.1 | 3 - 4 | 1061 | 112 | ||
GenBank mRNA | FQ211521.1 | - | 3 - 4 | 853 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
