Assay Details
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107863.1 | NP_001101333.1 | 3 - 4 | 832 | 117 | |
XM_006236571.2 | XP_006236633.1 | 4 - 5 | 1163 | 117 | ||
GenBank mRNA | FQ211521.1 | - | 4 - 5 | 955 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
