Assay Details
Gene Symbol: Crhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022714.1 | NP_073205.1 | 7 - 8 | 973 | 64 | |
XM_006236556.2 | XP_006236618.1 | 8 - 9 | 963 | 64 | ||
XM_006236557.2 | XP_006236619.1 | 8 - 9 | 960 | 64 | ||
XM_006236558.1 | XP_006236620.1 | 7 - 8 | 991 | 64 | ||
GenBank mRNA | U16253.1 | - | 7 - 8 | 973 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
