Assay Details
Gene Symbol: Npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012614.2 | NP_036746.1 | 3 - 4 | 372 | 130 | |
GenBank mRNA | JX157883.1 | - | 3 - 4 | 193 | 130 | |
M15880.1 | - | 3 - 4 | 347 | 130 | ||
M20373.1 | - | 3 - 4 | 327 | 130 |
Gene Symbol: LOC100912228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749745.3 | XP_003749793.1 | 385 | 130 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
