Assay Details
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126296.2 | NP_001119768.2 | 2 - 3 | 336 | 94 | |
XM_006236464.2 | XP_006236526.1 | 2 - 3 | 355 | 94 | ||
XM_006236465.2 | XP_006236527.1 | 2 - 3 | 355 | 94 | ||
XM_006236466.2 | XP_006236528.1 | 2 - 3 | 355 | 94 | ||
GenBank mRNA | BC161978.1 | - | 2 - 3 | 336 | 9029930 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
