Assay Details
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126296.2 | NP_001119768.2 | 3 - 4 | 493 | 121 | |
XM_006236464.2 | XP_006236526.1 | 3 - 4 | 512 | 121 | ||
XM_006236465.2 | XP_006236527.1 | 3 - 4 | 512 | 121 | ||
XM_006236466.2 | XP_006236528.1 | 3 - 4 | 512 | 121 | ||
GenBank mRNA | BC161978.1 | - | 3 - 4 | 493 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
