Assay Details
Gene Symbol: Hnrnpa2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104613.1 | NP_001098083.1 | 7 - 8 | 757 | 141 | |
GenBank mRNA | AB006815.1 | - | 6 - 7 | 721 | 141 | |
AB006816.1 | - | 7 - 8 | 757 | 141 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
