Assay Details
Gene Symbol: Actr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191860.1 | NP_001178789.1 | 1013 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
