Assay Details
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013209.1 | NP_001013227.1 | 2 - 3 | 188 | 79 | |
XM_006235863.2 | XP_006235925.1 | 2 - 3 | 234 | 79 | ||
XM_006235864.2 | XP_006235926.1 | 3 - 4 | 291 | 79 | ||
XM_006235866.2 | XP_006235928.1 | 2 - 3 | 231 | 79 | ||
GenBank mRNA | BC078908.1 | - | 2 - 3 | 188 | 7113862 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
