Assay Details
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030990.2 | NP_112252.1 | 4 - 5 | 771 | 106 | |
XM_006257293.2 | XP_006257355.1 | 7 - 8 | 1053 | 106 | ||
XM_008773401.1 | XP_008771623.1 | 7 - 8 | 1157 | 106 | ||
GenBank mRNA | BC093596.1 | - | 3 - 4 | 632 | 7314758 | 106 |
FQ212519.1 | - | 3 - 4 | 380 | 106 | ||
FQ212680.1 | - | 3 - 4 | 485 | 106 | ||
FQ213540.1 | - | 3 - 4 | 485 | 106 | ||
FQ213943.1 | - | 3 - 4 | 485 | 106 | ||
FQ214012.1 | - | 3 - 4 | 485 | 106 | ||
FQ214050.1 | - | 3 - 4 | 485 | 106 | ||
FQ214556.1 | - | 3 - 4 | 485 | 106 | ||
M11185.1 | - | 4 - 5 | 746 | 106 | ||
M14126.1 | - | 1 - 2 | 99 | 106 | ||
M25888.1 | - | 2 - 3 | 311 | 106 | ||
X02809.1 | - | 4 - 5 | 675 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
