Assay Details
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030990.2 | NP_112252.1 | 1 - 2 | 149 | 62 | |
XM_006257293.2 | XP_006257355.1 | 4 - 5 | 536 | 62 | ||
XM_008773401.1 | XP_008771623.1 | 4 - 5 | 535 | 62 | ||
GenBank mRNA | CB697155.1 | - | 1 - 2 | 160 | 62 | |
FQ211530.1 | - | 1 - 2 | 164 | 62 | ||
M11185.1 | - | 1 - 2 | 124 | 62 | ||
X02809.1 | - | 1 - 2 | 53 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M11185.1, NM_030990.2, X02809.1, XM_006257293.2, XM_008773401.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
