Assay Details
Gene Symbol: Il1rapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166342.1 | NP_001159814.1 | 3 - 4 | 1278 | 69 | |
XM_008773406.1 | XP_008771628.1 | 2 - 3 | 543 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
