Assay Details
Gene Symbol: As3mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080890.1 | NP_543166.1 | 5 - 6 | 497 | 90 | |
GenBank mRNA | AF393243.1 | - | 5 - 6 | 497 | 90 | |
BC061533.1 | - | 5 - 6 | 521 | 6918024 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
