Assay Details
Gene Symbol: As3mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080890.1 | NP_543166.1 | 7 - 8 | 649 | 89 | |
GenBank mRNA | AF393243.1 | - | 7 - 8 | 649 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
