Assay Details
Gene Symbol: Cnnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107593.1 | NP_001101063.1 | 2 - 3 | 950 | 96 | |
XM_006231483.2 | XP_006231545.1 | 2 - 3 | 1849 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
