Assay Details
Gene Symbol: Cnnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107593.1 | NP_001101063.1 | 4 - 5 | 1260 | 111 | |
XM_006231483.2 | XP_006231545.1 | 4 - 5 | 2159 | 111 | ||
XM_008760426.1 | XP_008758648.1 | 3 - 4 | 2018 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
