Assay Details
Gene Symbol: Cnnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107593.1 | NP_001101063.1 | 8 - 9 | 1750 | 74 | |
XM_006231483.2 | XP_006231545.1 | 9 - 10 | 2712 | 74 | ||
XM_008760426.1 | XP_008758648.1 | 8 - 9 | 2571 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
