Assay Details
Gene Symbol: Cnnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011942.1 | NP_001011942.1 | 5 - 6 | 2275 | 109 | |
GenBank mRNA | BC085930.1 | - | 5 - 6 | 2275 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P494439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156796 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P494439 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P494439 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P494439 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P494439 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84484 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83073 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22428 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100441 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84112 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53346 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26957 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53971 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10303 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10715891 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715898 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715892 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715896 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715893 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715895 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715894 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715897 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10715889 | - | - |
Rat Genome 230 2.0 Array | 1380970_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380970_at | - | - |
Rat Genome 230 2.0 Array | 1382269_at | - | - |
Rat Genome 230 2.0 Array | 1382269_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382269_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382269_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100030685.rn.1 | TC0100003757.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100030686.rn.1 | TC0100003757.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100030684.rn.1 | TC0100003757.rn.1 | RTA TC |
Rat Expression Set 230 | 1382269_PM_at | - | - |
Rat Expression Set 230 | 1382269_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715889 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22425604 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375530 | - | - |
