Assay Details
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231604.1 | XP_006231666.1 | 1 - 2 | 683 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
