Assay Details
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022700.2 | NP_073191.1 | 1 - 2 | 152 | 137 | |
XM_006231559.2 | XP_006231621.1 | 2 - 3 | 673 | 137 | ||
GenBank mRNA | BC084722.1 | - | 1 - 2 | 152 | 137 | |
U12568.1 | - | 1 - 2 | 156 | 137 | ||
X76921.1 | - | 2 - 3 | 284 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
