Assay Details
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760547.1 | XP_008758769.1 | 1 - 2 | 1085 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
