Assay Details
Gene Symbol: Gsto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012071.1 | NP_001012071.1 | 3 - 4 | 530 | 108 | |
XM_006231590.2 | XP_006231652.1 | 3 - 4 | 492 | 108 | ||
XM_006231591.2 | XP_006231653.1 | 3 - 4 | 464 | 108 | ||
XM_006231592.1 | XP_006231654.1 | 3 - 4 | 1222 | 108 | ||
GenBank mRNA | BC079295.1 | - | 3 - 4 | 530 | 7134485 | 108 |
Gene Symbol: LOC103690044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760499.1 | XP_008758721.1 | 1 - 2 | 1199 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC079295.1, NM_001012071.1, XM_006231590.2, XM_006231591.2, XM_006231592.1, XM_008760499.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
