Assay Details
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014168.1 | NP_001014190.1 | 8 - 9 | 1405 | 80 | |
XM_006231545.2 | XP_006231607.1 | 7 - 8 | 1311 | 80 | ||
GenBank mRNA | BC085841.1 | - | 8 - 9 | 1405 | 7135812 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
